2-Bromo-5-methoxybenzoic acid, 98+%
2-Bromo-5-methoxybenzoic acid, 98+%
2-Bromo-5-methoxybenzoic acid, 98+%
Thermo Scientific Chemicals

2-Bromo-5-methoxybenzoic acid, 98+%

CAS: 22921-68-2 | C8H7BrO3 | 231.045 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12261.09
also known as A12261-09
Price (USD)
56.65
Online Exclusive
63.30
Save 6.65 (11%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
63.30
Save 6.65 (11%)
Each
Chemical Identifiers
CAS22921-68-2
IUPAC Name2-bromo-5-methoxybenzoate
Molecular FormulaC8H6BrO3
InChI KeyODHJOROUCITYNF-UHFFFAOYSA-M
SMILESCOC1=CC=C(Br)C(=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)>98.0%
Assay (Aqueous acid-base Titration)>98.0 to <102.0%
Appearance (Color)White to cream
FormCrystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only