1,3,5-Trichloro-2-nitrobenzene, 98+%
1,3,5-Trichloro-2-nitrobenzene, 98+%
1,3,5-Trichloro-2-nitrobenzene, 98+%
Thermo Scientific Chemicals

1,3,5-Trichloro-2-nitrobenzene, 98+%

CAS: 18708-70-8 | C6H2Cl3NO2 | 226.437 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12262.22
also known as A12262-22
Price (USD)
321.65
Special offer
Online exclusive
Ends: 30-Jun-2026
378.00
Save 56.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
321.65
Special offer
Online exclusive
Ends: 30-Jun-2026
378.00
Save 56.35 (15%)
Each
Chemical Identifiers
CAS18708-70-8
IUPAC Name1,3,5-trichloro-2-nitrobenzene
Molecular FormulaC6H2Cl3NO2
InChI KeyAEBJDOTVYMITIA-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=C(Cl)C=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow
Melting Point (clear melt)67.0-73.0?C
FormCrystals or powder or crystalline powder or lumps
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only