2-Fluoro-3-methylbenzoic acid, 98%
2-Fluoro-3-methylbenzoic acid, 98%
2-Fluoro-3-methylbenzoic acid, 98%
2-Fluoro-3-methylbenzoic acid, 98%
Thermo Scientific Chemicals

2-Fluoro-3-methylbenzoic acid, 98%

CAS: 315-31-1 | C8H7FO2 | 154.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12280.03
also known as A12280-03
Price (USD)
24.60
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
24.60
Each
Add to cart
Chemical Identifiers
CAS315-31-1
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Silylated GC)≥97.5%
Appearance (Color)Pale cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)115.0-121.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only