2-Aminobenzonitrile, 98%
2-Aminobenzonitrile, 98%
2-Aminobenzonitrile, 98%
Thermo Scientific Chemicals

2-Aminobenzonitrile, 98%

CAS: 1885-29-6 | C7H6N2 | 118.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A12286.30
also known as A12286-30
Price (USD)
193.65
Online Exclusive
215.00
Save 21.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
193.65
Online Exclusive
215.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS1885-29-6
IUPAC Name2-aminobenzonitrile
Molecular FormulaC7H6N2
InChI KeyHLCPWBZNUKCSBN-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystalline powder, flakes, prills or chunks
Appearance (Color)White to cream or pale yellow/yellow to pale brown
Assay (GC)≥97.5%
Melting Point (clear melt)46-52?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only