Benzo[b]thiophene-2-carboxamide, 97%
Benzo[b]thiophene-2-carboxamide, 97%
Benzo[b]thiophene-2-carboxamide, 97%
Benzo[b]thiophene-2-carboxamide, 97%
Thermo Scientific Chemicals

Benzo[b]thiophene-2-carboxamide, 97%

CAS: 6314-42-7 | C9H7NOS | 177.221 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12292.06
also known as A12292-06
Price (USD)
178.65
Special offer
Online exclusive
Ends: 30-Jun-2026
210.00
Save 31.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
178.65
Special offer
Online exclusive
Ends: 30-Jun-2026
210.00
Save 31.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS6314-42-7
IUPAC Name1-benzothiophene-2-carboxamide
Molecular FormulaC9H7NOS
InChI KeyGYSCBCSGKXNZRH-UHFFFAOYSA-N
SMILESNC(=O)C1=CC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Cream to pale brown
Melting Point (clear melt)170.5-179.5?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only