3-Methylcatechol, 97%
3-Methylcatechol, 97%
3-Methylcatechol, 97%
Thermo Scientific Chemicals

3-Methylcatechol, 97%

CAS: 488-17-5 | C7H8O2 | 124.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12324.14
also known as A12324-14
Price (USD)
116.65
Online Exclusive
129.00
Save 12.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
116.65
Online Exclusive
129.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS488-17-5
IUPAC Name3-methylbenzene-1,2-diol
Molecular FormulaC7H8O2
InChI KeyPGSWEKYNAOWQDF-UHFFFAOYSA-N
SMILESCC1=CC=CC(O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Melting Point (clear melt)60.0-69.0?C
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or fused solid or lumps
Used in organic synthesis. Used for the synthesis of antibacterial agent, antioxidants, As a polymer inhibitor, stabilizer, flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in organic synthesis. Used for the synthesis of antibacterial agent, antioxidants, As a polymer inhibitor, stabilizer, flavors.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Leonie E Hüskena,; Marcella C.F Dalma,; Johannes Trampera,; Jan Wery1,; Jan A.M de Bont,; Rik Beeftink. Integrated bioproduction and extraction of 3-methylcatechol. Journal of Biotechnology. 2001, 88 (1),11-19.
  2. Leonie E. Hüskena,; Mirjam Oomesa,; Karin Schroëna,; Johannes Trampera,; Jan A.M. de Bontc,; Rik Beeftink.Membrane-facilitated bioproduction of 3-methylcatechol in an octanol/water two-phase systemJournal of Biotechnology. 2002 , 96(3),281-289.