Sodium nitrate, 98+%
Sodium nitrate, 98+%
Sodium nitrate, 98+%
Thermo Scientific Chemicals

Sodium nitrate, 98+%

CAS: 7631-99-4 ∣ NNaO3 ∣ 84.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12327.0E
also known as A12327-0E
Price (USD)
99.65
Online exclusive
111.00
Save 11.35
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
99.65
Online exclusive
111.00
Save 11.35
Each
Chemical Identifiers
CAS7631-99-4
IUPAC Namesodium nitrate
Molecular FormulaNNaO3
InChI KeyVWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES[Na+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline powder or prills
Assay (unspecified)≥98.0%
CommentAs a bulk commodity material, may contain a small quantity of dark coloured prills from the manufacturing process
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications

Sodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.

Solubility

Freely soluble in water, ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine.

Notes

Sodium nitrate is hygroscopic in nature.

RUO – Research Use Only

General References:

  1. Xu, M.; Wang, J.; Li, C.; Zhang, J.; Liu, Y.; Yuan, G. Effect of electric field to infrared absorption properties of sodium nitrate on silver/diamond powder (Ag/DP) composites. Chem. Phys. Lett. 2014, 613, 10-14.
  2. Steinitz, M. O.; Pink, D. A.; Clancy, J. P.; MacDonald, N.; Swainson, I. Sodium Nitrate -- A Difficult Discontinuous Phase Transition. Can. J. Phys. 2004, 82 (12), 1097-1107.
  3. Hernández-Paredesa, J.; Carrillo-Pesqueiraa, F. J.; Esparza-Ponceb, H. E.; Hernández-Negretea, O.; Alvarez-Ramosa, M. E. l-Proline-sodium nitrate obtained from solvent drop grinding. Polyhedron 2015, 91, 84-88.