1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%
1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%
1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%
Thermo Scientific Chemicals

1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%

CAS: 328-70-1 | C8H3BrF6 | 293.006 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12337.09
also known as A12337-09
Price (USD)
39.65
Online Exclusive
43.40
Save 3.75 (9%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.40
Save 3.75 (9%)
Each
Chemical Identifiers
CAS328-70-1
IUPAC Name1-bromo-3,5-bis(trifluoromethyl)benzene
Molecular FormulaC8H3BrF6
InChI KeyCSVCVIHEBDJTCJ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(=CC(Br)=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Refractive Index1.4255-1.4295 @ 20?C
FormLiquid
Assay (GC)≥97.5%
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Guerrand, H. D. S.; Vaultier, M.; Pinet, S.; Pucheault, M. Amine-Borane Complexes: Air- and Moisture-Stable Partners for Palladium-Catalyzed Borylation of Aryl Bromides and Chlorides. Adv. Synth. Catal. 2015, 357 (6), 1167-1174.
  2. Soni, V.; Patel, U. N.; Punji, B. Metal-free regioselective C-3 acetoxylation of N-substituted indoles: crucial impact of nitrogen-substituent. RSC Adv. 2015, 5 (71), 57472-57481.