1-Bromo-2-chloroethane, 98%
1-Bromo-2-chloroethane, 98%
1-Bromo-2-chloroethane, 98%
Thermo Scientific Chemicals

1-Bromo-2-chloroethane, 98%

CAS: 107-04-0 | C2H4BrCl | 143.408 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A12349.0E
also known as A12349-0E
Price (USD)
825.65
Online Exclusive
917.00
Save 91.35 (10%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
825.65
Online Exclusive
917.00
Save 91.35 (10%)
Each
Chemical Identifiers
CAS107-04-0
IUPAC Name1-bromo-2-chloroethane
Molecular FormulaC2H4BrCl
InChI KeyIBYHHJPAARCAIE-UHFFFAOYSA-N
SMILESClCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4880-1.4920 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

Solubility
Soluble in water ( 7 g/L (20°C). Soluble in ethanol, ethyl ether, chloroform.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Yan Yang et. al. Dissociative double ionization of 1-bromo-2-chloroethane irradiated by an intense femtosecond laser field. Journal of Chemical Physics. 2001, 66 (18),6057-6063.
  2. Haiyan Wang,; Yining Huang.A conformational study of 1-bromo-2-chloroethane in zeolites. Langmuir. 2009, 25(14),8042-8050.