1,10-Dibromodecane, 97%
1,10-Dibromodecane, 97%
1,10-Dibromodecane, 97%
Thermo Scientific Chemicals

1,10-Dibromodecane, 97%

CAS: 4101-68-2 | C10H20Br2 | 300.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A12357.36
also known as A12357-36
Price (USD)
356.65
Online Exclusive
396.00
Save 39.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
356.65
Online Exclusive
396.00
Save 39.35 (10%)
Each
Chemical Identifiers
CAS4101-68-2
IUPAC Name1,10-dibromodecane
Molecular FormulaC10H20Br2
InChI KeyGTQHJCOHNAFHRE-UHFFFAOYSA-N
SMILESBrCCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)23.0-33.0?C
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sea
RUO – Research Use Only

General References:

  1. V. Percec.; M. Kawasumi.; P. L. Rinaldi.; V. E. Litman. Synthesis and characterization of cyclic liquid crystalline oligomers based on 1-(4-hydroxy-4'-biphenylyl)-2-(4-hydroxyphenyl)butane and 1,10-dibromodecane. Macromolecules. 1992, 25 (15), 3851-3861.
  2. Abil E. Aliev.; Sharon P. Smart.; Ian J. Shannon.; Kenneth D. M. Harris. Structural and dynamic properties of the 1,10-dibromodecane/urea inclusion compound, investigated by variable-temperature powder X-ray diffraction, solid-state 2H NMR lineshape analysis and solid-state 2H NMR spin-lattice relaxation time measurements. J. Chem. Soc., Faraday Trans. 1992, 96 (12), 2179-2185.