Complexometric Titration≥97.5 to ≤102.5% (Dry wt. basis)
Water Content (Karl Fischer Titration)<10%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications Calcium benzoate hydrate is used in used in the food industry as a preservative.
Solubility Soluble in water (2.6 g/100ml at 0°C).
Notes Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.