Thermo Scientific Chemicals

4-Chlorobenzoylacetonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A12364.22
100 g, Each
Thermo Scientific Chemicals

4-Chlorobenzoylacetonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A12364.22
100 g, Each
Quantity
Catalog number: A12364.22
also known as A12364-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
4640-66-8
IUPAC Name
3-(4-chlorophenyl)-3-oxopropanenitrile
Molecular Formula
C9H6ClNO
InChI Key
JYOUFPNYTOFCSJ-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)C(=O)CC#N
Melting Point (clear melt)
126.0-133.0?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale cream to cream to yellow to orange
Assay (GC)
≥97.5%

Description

Pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical Intermediates.

Solubility
Soluble in Dichloromethane, Ether, Ethyl Acetate and Methanol

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text