Ethyl (ethoxymethylene)cyanoacetate, 98%
Ethyl (ethoxymethylene)cyanoacetate, 98%
Ethyl (ethoxymethylene)cyanoacetate, 98%
Ethyl (ethoxymethylene)cyanoacetate, 98%
Thermo Scientific Chemicals

Ethyl (ethoxymethylene)cyanoacetate, 98%

CAS: 94-05-3 | C8H11NO3 | 169.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12366.18
also known as A12366-18
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.60
Save 5.95 (15%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.60
Save 5.95 (15%)
Each
Add to cart
Chemical Identifiers
CAS94-05-3
IUPAC Nameethyl (2Z)-2-cyano-3-ethoxyprop-2-enoate
Molecular FormulaC8H11NO3
InChI KeyKTMGNAIGXYODKQ-SREVYHEPSA-N
SMILESCCO\C=C(\C#N)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid or flakes
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Appearance (Color)White to pale cream to cream to yellow to brown
Melting Point (clear melt)47.0-53.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only