Pentafluorobenzoic acid, 98+%
Pentafluorobenzoic acid, 98+%
Pentafluorobenzoic acid, 98+%
Thermo Scientific Chemicals

Pentafluorobenzoic acid, 98+%

CAS: 602-94-8 | C7HF5O2 | 212.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12396.14
also known as A12396-14
Price (USD)
134.65
Online Exclusive
149.00
Save 14.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
134.65
Online Exclusive
149.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS602-94-8
IUPAC Namepentafluorobenzoic acid
Molecular FormulaC7HF5O2
InChI KeyYZERDTREOUSUHF-UHFFFAOYSA-N
SMILESOC(=O)C1=C(F)C(F)=C(F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Identification (FTIR)Conforms
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only