Ethyl 3-bromopropionate, 99%
Ethyl 3-bromopropionate, 99%
Ethyl 3-bromopropionate, 99%
Ethyl 3-bromopropionate, 99%
Thermo Scientific Chemicals

Ethyl 3-bromopropionate, 99%

CAS: 539-74-2 | C5H9BrO2 | 181.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12402.36
also known as A12402-36
Price (USD)
816.65
Online Exclusive
907.00
Save 90.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
816.65
Online Exclusive
907.00
Save 90.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS539-74-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.4510-1.4540 @ 20°C
FormLiquid
Ethyl 3-bromopropionate was used in the preparation of (3?,4?-dimethoxyphenyl)-5-oxopentanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-bromopropionate was used in the preparation of (3′,4′-dimethoxyphenyl)-5-oxopentanoic acid.

Solubility
Soluble in water, alcohol, chloroform, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Bases, Reducing agents
RUO – Research Use Only

General References:

  1. Berlin KD,; Herd MD. Proc. Novel 2-amino-4-aryl-substituted-and 2-amino-4, 5-disubstituted-thiazoles.Oklahoma Acad. Sci. 1991, 71 29-33
  2. Undergoes a homo-Reformatsky coupling with carbonyl compounds leading to -lactones. The homoenolate can be generated with various lanthanide metals, La, Ce, Nd or Sm: J. Org. Chem., 55, 1628 (1990):
  3. Cyclopropanols may be synthesized by reaction of ß-halo esters with Grignard reagents and SmI2: Tetrahedron Lett., 32, 7691 (1991).