Taurine, 99%
Taurine, 99%
Taurine, 99%
Thermo Scientific Chemicals

Taurine, 99%

CAS: 107-35-7 | C2H7NO3S | 125.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A12403.36
also known as A12403-36
Price (USD)
140.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
140.00
Each
Chemical Identifiers
CAS107-35-7
IUPAC Name2-aminoethane-1-sulfonic acid
Molecular FormulaC2H7NO3S
InChI KeyXOAAWQZATWQOTB-UHFFFAOYSA-N
SMILESNCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder or needles
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
Assay from Suppliers CofA≥98.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Appearance (Color)White
View more
Taurine plays an important role in cardiovascular function, skeletal muscle function, the retina and the central nervous system. It acts as an antioxidant as well as protecting against the toxicity of various substances such as lead and cadmium. It is also used in contact lens solution and an ingredient in numerous energy drinks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Taurine plays an important role in cardiovascular function, skeletal muscle function, the retina and the central nervous system. It acts as an antioxidant as well as protecting against the toxicity of various substances such as lead and cadmium. It is also used in contact lens solution and an ingredient in numerous energy drinks.

Solubility
Soluble in water and dimethyl sulfoxide. Insoluble in absolute ethanol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. For a monograph, see: Taurine, R. Huxtable, A. Barbeau, Eds., Raven Press, N.Y., (1975).
  2. Jurkowska, H.; Stipanuk, M. H.; Hirschberger, L. L.; Roman, H. B. Propargylglycine inhibits hypotaurine/taurine synthesis and elevates cystathionine and homocysteine concentrations in primary mouse hepatocytes. Amino acids 2015, 47 (6), 1215-1223.
  3. Chou, C. T.; Lin, H. T.; Hwang, P. A.; Wang, S. T.; Hsieh, C. H.; Hwang, D. F. Taurine resumed neuronal differentiation in arsenite-treated N2a cells through reducing oxidative stress, endoplasmic reticulum stress, and mitochondrial dysfunction. Amino acids 2015, 47 (4), 735-744.