1-Fluoro-2-iodobenzene, 99%, stab. with copper
1-Fluoro-2-iodobenzene, 99%, stab. with copper
1-Fluoro-2-iodobenzene, 99%, stab. with copper
Thermo Scientific Chemicals

1-Fluoro-2-iodobenzene, 99%, stab. with copper

CAS: 348-52-7 | C6H4FI | 222.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12411.22
also known as A12411-22
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS348-52-7
IUPAC Name1-fluoro-2-iodobenzene
Molecular FormulaC6H4FI
InChI KeyTYHUGKGZNOULKD-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Refractive Index1.5880 - 1.5920 @20?C
Assay (GC)> 98.5%
Appearance (Color)Clear, colourless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only