2,4-Difluorophenyl isocyanate, 98+%
2,4-Difluorophenyl isocyanate, 98+%
2,4-Difluorophenyl isocyanate, 98+%
Thermo Scientific Chemicals

2,4-Difluorophenyl isocyanate, 98+%

CAS: 59025-55-7 | C7H3F2NO | 155.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12415.14
also known as A12415-14
Price (USD)
135.65
Online Exclusive
150.00
Save 14.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
135.65
Online Exclusive
150.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS59025-55-7
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormLiquid
Refractive Index1.4905-1.4945 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only