4-Isopropylphenylhydrazine hydrochloride, 98%
4-Isopropylphenylhydrazine hydrochloride, 98%
4-Isopropylphenylhydrazine hydrochloride, 98%
Thermo Scientific Chemicals

4-Isopropylphenylhydrazine hydrochloride, 98%

CAS: 118427-29-5 | C9H14N2 | 150.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12416.03
also known as A12416-03
Price (USD)
35.50
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.50
Each
Chemical Identifiers
CAS118427-29-5
IUPAC Name[4-(propan-2-yl)phenyl]hydrazine
Molecular FormulaC9H14N2
InChI KeyZYATZFJUOXJFPY-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(NN)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown or yellow
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
FormPowder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only