Ethyl 4-pentenoate, 98+%
Ethyl 4-pentenoate, 98+%
Ethyl 4-pentenoate, 98+%
Thermo Scientific Chemicals

Ethyl 4-pentenoate, 98+%

CAS: 1968-40-7 | C7H12O2 | 128.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Catalog number A12422.03
also known as A12422-03
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.80
Save 5.15
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.80
Save 5.15
Each
Chemical Identifiers
CAS1968-40-7
IUPAC Nameethyl pent-4-enoate
Molecular FormulaC7H12O2
InChI KeyPTVSRINJXWDIKP-UHFFFAOYSA-N
SMILESCCOC(=O)CCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.4135-1.4175 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Ethyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Das, D. D.; McEnally, C. S.; Pfefferle, L. D. Sooting tendencies of unsaturated esters in nonpremixed flames. Combust. Flame 2015, 162 (4), 1489-1497.
  2. Tellitu, I.; Beitia, I.; Díaz, M.; Alonso, A.; Moreno, I.; Domínguez, E. An improved solvent-free system for the microwave-assisted decarboxylation of malonate derivatives based on the use of imidazole. Tetrahedron 2015, 71 (43), 8251-8255.