2-Benzyloxybenzoic acid, 98%
2-Benzyloxybenzoic acid, 98%
2-Benzyloxybenzoic acid, 98%
Thermo Scientific Chemicals

2-Benzyloxybenzoic acid, 98%

CAS: 14389-86-7 | C14H11O3 | 227.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12425.30
also known as A12425-30
Price (USD)
1,052.65
Online Exclusive
1,169.00
Save 116.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
1,052.65
Online Exclusive
1,169.00
Save 116.35 (10%)
Each
Chemical Identifiers
CAS14389-86-7
IUPAC Name2-(benzyloxy)benzoate
Molecular FormulaC14H11O3
InChI KeyGMOYUTKNPLBTMT-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=CC=C1OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)72.5-78.5°C
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only