4-(Methylamino)benzoic acid, 97%
4-(Methylamino)benzoic acid, 97%
4-(Methylamino)benzoic acid, 97%
Thermo Scientific Chemicals

4-(Methylamino)benzoic acid, 97%

CAS: 10541-83-0 | C8H9NO2 | 151.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12426.14
also known as A12426-14
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
127.00
Save 19.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
127.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS10541-83-0
IUPAC Name4-(methylamino)benzoic acid
Molecular FormulaC8H9NO2
InChI KeyZVIDMSBTYRSMAR-UHFFFAOYSA-N
SMILESCNC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)156.0-166.0?C
Appearance (Color)Pale cream to cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-(Methylamino)benzoic acid important for the preparation of other pharmaceutical products, dyes, flavors, and preservatives. .For the preparation of medicines and other fine chemicals.

Solubility
Slightly soluble. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Farook Adam,; Jeyashelly Andas.Amino benzoic acid modified silica--an improved catalyst for the mono-substituted product in the benzylation of toluene with benzyl chloride.. Journal of Colloid and Interface Science.2007, 311 (1), 135-143.
  2. Andrea Martorana et. al. Insights into the homocoupling reaction of 4-methylamino benzoic acid mediated by Trametes versicolor laccase.. Molecular BioSystems.2011, 7 (11) , 2967-2969 .