Cyclobutanol, 96%
Cyclobutanol, 96%
Cyclobutanol, 96%
Thermo Scientific Chemicals

Cyclobutanol, 96%

CAS: 2919-23-5 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A12430.03
also known as A12430-03
Price (USD)
105.65
Special offer
Online exclusive
Ends: 30-Jun-2026
124.00
Save 18.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
105.65
Special offer
Online exclusive
Ends: 30-Jun-2026
124.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS2919-23-5
IUPAC Namecyclobutanol
Molecular FormulaC4H8O
InChI KeyKTHXBEHDVMTNOH-UHFFFAOYSA-N
SMILESOC1CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4335-1.4395 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥95.0%
Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.

Solubility
Partly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid chlorides, acid anhydrides.
RUO – Research Use Only

General References:

  1. Jan. Rocek,; Annette E. Radkowsky.Mechanism of the chromic acid oxidation of cyclobutanol. J. Am. Chem. Soc. 1973 , 95(21),7123-7132.
  2. Jan Rocek,; Annette E. Radkowsky.The role of chromium(IV) in oxidations by chromic acid. The oxidative cleavage of cyclobutanol. J. Am. Chem. Soc.1968 , 90(11), 2986-2988.