2,4,6-Tribromoaniline, 98%
2,4,6-Tribromoaniline, 98%
2,4,6-Tribromoaniline, 98%
Thermo Scientific Chemicals

2,4,6-Tribromoaniline, 98%

CAS: 147-82-0 | C6H4Br3N | 329.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A12453.36
also known as A12453-36
Price (USD)
194.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
194.00
Each
Chemical Identifiers
CAS147-82-0
IUPAC Name2,4,6-tribromoaniline
Molecular FormulaC6H4Br3N
InChI KeyGVPODVKBTHCGFU-UHFFFAOYSA-N
SMILESNC1=C(Br)C=C(Br)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormPowder
Melting Point (clear melt)116.0-123.0°C
Appearance (Color)White to pale cream to brown or pale purple to gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only