3,4-Difluoroanisole, 98%
3,4-Difluoroanisole, 98%
3,4-Difluoroanisole, 98%
Thermo Scientific Chemicals

3,4-Difluoroanisole, 98%

CAS: 115144-40-6 | C7H6F2O | 144.121 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12474.06
also known as A12474-06
Price (USD)
29.65
Special offer
Online exclusive
Ends: 28-Dec-2026
34.80
Save 5.15
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
29.65
Special offer
Online exclusive
Ends: 28-Dec-2026
34.80
Save 5.15
Each
Chemical Identifiers
CAS115144-40-6
IUPAC Name1,2-difluoro-4-methoxybenzene
Molecular FormulaC7H6F2O
InChI KeyDSEVNUCNUTYYHW-UHFFFAOYSA-N
SMILESCOC1=CC=C(F)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4660-1.4700 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only