o-Xylylene dichloride, 98%
o-Xylylene dichloride, 98%
o-Xylylene dichloride, 98%
o-Xylylene dichloride, 98%
Thermo Scientific Chemicals

o-Xylylene dichloride, 98%

CAS: 612-12-4 | C8H8Cl2 | 175.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A12500.30
also known as A12500-30
Price (USD)
221.65
Special offer
Online exclusive
Ends: 30-Jun-2026
261.00
Save 39.35 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
221.65
Special offer
Online exclusive
Ends: 30-Jun-2026
261.00
Save 39.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS612-12-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay (GC)≥97.5%
FormCrystalline powder or fused solid
Melting Point (clear melt)52.0-59.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only