Blue Tetrazolium chloride
Blue Tetrazolium chloride
Blue Tetrazolium chloride
Thermo Scientific Chemicals

Blue Tetrazolium chloride

CAS: 1871-22-3 | C40H36Cl2N8O2 | 731.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12502.06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1871-22-3
IUPAC Namedihydrogen 2-[4'-(3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazole dichloride
Molecular FormulaC40H36Cl2N8O2
InChI KeyRCEHREKDVGHYAM-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormPowder
Identification (FTIR)Conforms
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Blue Tetrazolium chloride is useful in the prepartion of DNA cation complexes and immunoadsorbents for anti-DNA antibodies. It is used in assay of reducing sugars. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Blue Tetrazolium chloride is useful in the prepartion of DNA cation complexes and immunoadsorbents for anti-DNA antibodies. It is used in assay of reducing sugars. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum.

Solubility
Soluble in hot water (5 mg/ml), methanol (50 mg/ml), ethanol, and chloroform. Insoluble in acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. C.K.Jue; P.N.Lipke. Determination of reducing sugars in the nanomole range with tetrazolium blue chloride. Journal of Biochemical and Biophysical Methods. 1985, 11, (2-3)109-115
  2. Vincent A.DeBari; Mark A.Needle. Cation complexed DNA. A novel class of immunoadsorbents for anti-DNA antibodies. Journal of Immunological Methods. 1981, 40, (1)89-94
  3. Reagent for identification of reducing sugars.
  4. For use in the oxidation of steroids, see: J. Chem. Soc., Chem. Commun., 2687 (1988).