3'-Aminoacetophenone, 97%
3'-Aminoacetophenone, 97%
3'-Aminoacetophenone, 97%
Thermo Scientific Chemicals

3'-Aminoacetophenone, 97%

CAS: 99-03-6 | C8H9NO | 135.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12503.0B
also known as A12503-0B
Price (USD)
253.65
Special offer
Online exclusive
Ends: 28-Dec-2026
298.00
Save 44.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
253.65
Special offer
Online exclusive
Ends: 28-Dec-2026
298.00
Save 44.35
Each
Chemical Identifiers
CAS99-03-6
IUPAC Name1-(3-aminophenyl)ethan-1-one
Molecular FormulaC8H9NO
InChI KeyCKQHAYFOPRIUOM-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream or yellow to brown
FormPowder or crystalline powder
Identification (FTIR)Conforms
Melting point95-100°C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only