3-Amino-5-nitrobenzoic acid, 98%
3-Amino-5-nitrobenzoic acid, 98%
3-Amino-5-nitrobenzoic acid, 98%
3-Amino-5-nitrobenzoic acid, 98%
Thermo Scientific Chemicals

3-Amino-5-nitrobenzoic acid, 98%

CAS: 618-84-8 | C7H6N2O4 | 182.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12508.06
also known as A12508-06
Price (USD)
92.65
Special offer
Online exclusive
Ends: 30-Jun-2026
109.00
Save 16.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 30-Jun-2026
109.00
Save 16.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS618-84-8
IUPAC Name3-amino-5-nitrobenzoic acid
Molecular FormulaC7H6N2O4
InChI KeyZNVHAQRPXAQKRU-UHFFFAOYSA-N
SMILESNC1=CC(=CC(=C1)[N+]([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to green to brown
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Melting Point (clear melt)214.0-220.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only