3-Acetylcoumarin, 98+%
3-Acetylcoumarin, 98+%
3-Acetylcoumarin, 98+%
Thermo Scientific Chemicals

3-Acetylcoumarin, 98+%

CAS: 3949-36-8 | C11H8O3 | 188.182 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12532.06
also known as A12532-06
Price (USD)
46.65
Online Exclusive
52.10
Save 5.45 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.65
Online Exclusive
52.10
Save 5.45 (10%)
Each
Chemical Identifiers
CAS3949-36-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
Assay (GC)≥98.0%
Melting Point (clear melt)120.0-126.0?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only