Ethyl 4-chlorobenzoate, 98+%
Ethyl 4-chlorobenzoate, 98+%
Ethyl 4-chlorobenzoate, 98+%
Ethyl 4-chlorobenzoate, 98+%
Thermo Scientific Chemicals

Ethyl 4-chlorobenzoate, 98+%

CAS: 7335-27-5 | C9H9ClO2 | 184.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A12540.30
also known as A12540-30
Price (USD)
198.65
Special offer
Online exclusive
Ends: 30-Jun-2026
233.00
Save 34.35 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
198.65
Special offer
Online exclusive
Ends: 30-Jun-2026
233.00
Save 34.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7335-27-5
IUPAC Nameethyl 4-chlorobenzoate
Molecular FormulaC9H9ClO2
InChI KeyRWBYCMPOFNRISR-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
Refractive Index1.5215-1.5255 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only