4-Nitrobenzoyl chloride, 98%
4-Nitrobenzoyl chloride, 98%
4-Nitrobenzoyl chloride, 98%
4-Nitrobenzoyl chloride, 98%
Thermo Scientific Chemicals

4-Nitrobenzoyl chloride, 98%

CAS: 122-04-3 | C7H4ClNO3 | 185.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
250 g
1000 g
Catalog number A12543.30
also known as A12543-30
Price (USD)
58.65
Online Exclusive
64.80
Save 6.15 (9%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
58.65
Online Exclusive
64.80
Save 6.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS122-04-3
IUPAC Name4-nitrobenzoyl chloride
Molecular FormulaC7H4ClNO3
InChI KeySKDHHIUENRGTHK-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystalline powder or crystals or flakes
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)70-75?C
4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.

Solubility
Soluble in terahydrofuran, dichloromethane, chloroform and pyridine.

Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Reagent for the preparation of crystalline esters.
  2. Chen, W.; Feng, Y.; Zhang, M.; Wu, J.; Zhang, J.; Gao, X.; He, J.; Zhang, J. Homogeneous benzoylation of cellulose in 1-allyl-3-methylimidazolium chloride: Hammett correlation, mechanism and regioselectivity. RSC Adv. 2015, 5 (72), 58536-58542.
  3. Skácel, J.; Budka, J.; Eigner, V.; Lhoták, P. Regioselective Friedel-Crafts acylation of calix[4]arenes. Tetrahedron 2015, 71 (13), 1959-1965.