2,4-Dimethoxybenzaldehyde, 98%
2,4-Dimethoxybenzaldehyde, 98%
2,4-Dimethoxybenzaldehyde, 98%
Thermo Scientific Chemicals

2,4-Dimethoxybenzaldehyde, 98%

CAS: 613-45-6 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12549.22
also known as A12549-22
Price (USD)
104.65
Special offer
Online exclusive
Ends: 30-Jun-2026
123.00
Save 18.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
104.65
Special offer
Online exclusive
Ends: 30-Jun-2026
123.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS613-45-6
IUPAC Name2,4-dimethoxybenzaldehyde
Molecular FormulaC9H10O3
InChI KeyLWRSYTXEQUUTKW-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)68-72?C
Appearance (Color)White to pale yellow or cream to pale brown
2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.

Solubility
Insoluble in water. Soluble in chlorofrom.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Aliakbar Dehno Khalaji; Karla Fejfarova; Michal Dusek. Synthesis and characterization of two Diimine Schiff Bases derived from 2,4-dimethoxybenzaldehyde: The crystal structure of N,N’-Bis(2,4-dimethoxybenzylidene)-1,2-diaminoethane. Acta Chim. Slov. 2010, 57, 257-261
  2. Kou-Chang Liu; Becky R.Shelton; Robert K.Howe. A particularly convenient preparation of benzohydroximinoyl chlorides (nitrile oxide precursors). J. Org. Chem. 1980, 45, (19)3916-3918
  3. Has been used to protect diols as their cyclic acetals, by azeotroping in the presence of TsOH: J. Am. Chem. Soc., 84, 430 (1962). The products are readily hydrolyzed by acid but are stable to hydrogenolysis conditions (Raney Ni) used to cleave O-benzyl groups: Tetrahedron, 42, 3021 (1986).