2'-Hydroxyacetophenone, 98%
2'-Hydroxyacetophenone, 98%
2'-Hydroxyacetophenone, 98%
Thermo Scientific Chemicals

2'-Hydroxyacetophenone, 98%

CAS: 118-93-4 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A12550.36
also known as A12550-36
Price (USD)
83.65
Online Exclusive
92.90
Save 9.25 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
83.65
Online Exclusive
92.90
Save 9.25 (10%)
Each
Chemical Identifiers
CAS118-93-4
IUPAC Name1-(2-hydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O2
InChI KeyJECYUBVRTQDVAT-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.5570-1.5610 @ 20?C
Appearance (Color)Clear colorless to pale yellow to yellow to orange
2'-Hydroxyacetophenone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxyacetophenone is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Andre Loupy; Serge Regnier. Solvent-free microwave-assisted Beckmann rearrangement of benzaldehyde and 2-hydroxyacetophenone oximes. Tetrahedron Letters. 1999, 40, (34)6221-6224
  2. Rohith P.John; A.Sreekanth; V.Rajakannan; T.A.Ajith; M.R.Prathapachandra Kurup. New copper(II) complexes of 2-hydroxyacetophenone N(4)-substituted thiosemicarbazones and polypyridyl co-ligands: structural, electrochemical and antimicrobial studies. Polyhedron. 2004, 23, (16)2549-2559