Dimethyl succinate, 98%
Dimethyl succinate, 98%
Dimethyl succinate, 98%
Thermo Scientific Chemicals

Dimethyl succinate, 98%

CAS: 106-65-0 | C6H10O4 | 146.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1000 g
Catalog number A12565.0B
also known as A12565-0B
Price (USD)
266.00
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
266.00
Each
Chemical Identifiers
CAS106-65-0
IUPAC Name1,4-dimethyl butanedioate
Molecular FormulaC6H10O4
InChI KeyMUXOBHXGJLMRAB-UHFFFAOYSA-N
SMILESCOC(=O)CCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5% (UK-sourced material)
Assay from Supplier's CofA≥97.5% (US-sourced material)
CommentMaterial sourced in UK and US
Identification (FTIR)Conforms (UK-sourced material)
View more
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.

Solubility
Soluble in water. (122,9 g/L at 20°C). soluble in acetone, ethanol, very soluble in ethyl ether.

Notes
Store at room temperature. Ensure good ventilation.
RUO – Research Use Only

General References:

  1. Susana Puertas.; Francisca Rebolledo.; Vicente Gotor. Selective ammonolysis and aminolysis of dimethyl succinate. Synthesis of optically active N-alkylsuccinimides. Tetrahedron. 1995, 51 (5),1495-1502.
  2. T. Tureka.; D.L. Trimm.; D.StC. Black.; N.W. Cant. Hydrogenolysis of dimethyl succinate on copperbased catalysts. Appl. Catal., A. 1994, 116 (1-2),137-150.