2-Aminobenzophenone, 98%
2-Aminobenzophenone, 98%
2-Aminobenzophenone, 98%
Thermo Scientific Chemicals

2-Aminobenzophenone, 98%

CAS: 2835-77-0 | C13H11NO | 197.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12580.09
also known as A12580-09
Price (USD)
39.65
Online Exclusive
44.20
Save 4.55 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
44.20
Save 4.55 (10%)
Each
Chemical Identifiers
CAS2835-77-0
IUPAC Name2-benzoylaniline
Molecular FormulaC13H11NO
InChI KeyMAOBFOXLCJIFLV-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)104.0-111.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow
Assay (GC)≥97.5%
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Insoluble in water. Soluble in DMSO, Methanol.

Notes
Store at room temperature. Ensure good ventilation.
RUO – Research Use Only

General References:

  1. Jing-Ping Liou.; Chun-Wei Chang.; Jeng-Shin Song.; Yung-Ning Yang.; Ching-Fang Yeh.; Huan-Yi Tseng.; Yu-Kang Lo.; Yi-Ling Chang.; Chung-Ming Chang.; Hsing-Pang Hsieh. Synthesis and Structure-Activity Relationship of 2-Aminobenzophenone Derivatives as Antimitotic Agents. J. Med. Chem. 2002, 45 (12), 2556-2562.
  2. Yuri N. Belokon.; Vitali I. Tararov.; Viktor I. Maleev.; Tatiana F. Savel'eva.; Michael G. Ryzhov. Improved procedures for the synthesis of (S)-2-[N-(N'-benzylprolyl)amino]benzophenone (BPB) and Ni(II) complexes of Schiff's bases derived from BPB and amino acids. Tetrahedron: Asymmetry. 1998, 9 (23), 4249-4252.