Diphenyl disulfide, 98%
Diphenyl disulfide, 98%
Diphenyl disulfide, 98%
Thermo Scientific Chemicals

Diphenyl disulfide, 98%

CAS: 882-33-7 | C12H10S2 | 218.332 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A12586.18
also known as A12586-18
Price (USD)
51.65
Online exclusive
57.50
Save 5.85
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
51.65
Online exclusive
57.50
Save 5.85
Each
Chemical Identifiers
CAS882-33-7
IUPAC Name(phenyldisulfanyl)benzene
Molecular FormulaC12H10S2
InChI KeyGUUVPOWQJOLRAS-UHFFFAOYSA-N
SMILESS(SC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)57.0-63.0?C
Diphenyl disulfide is one of the most popular organic disulfides used in organic synthesis. It is used as a reagent for the α-phenylsulfenylation of carbonyl compounds. It participates in hydrothiolation of alkynes via amine-mediated single electron transfer mechanism. It is the hydrolysis product of dyfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl disulfide is one of the most popular organic disulfides used in organic synthesis. It is used as a reagent for the α-phenylsulfenylation of carbonyl compounds. It participates in hydrothiolation of alkynes via amine-mediated single electron transfer mechanism. It is the hydrolysis product of dyfonate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Tsuyoshi Taniguchi; Tatsuya Fujii; Atsushi Idota; Hiroyuki Ishibashi. Reductive addition of the benzenethiyl radical to alkynes by amine-mediated single electron transfer reaction to diphenyl disulfide. Organic Letters. 2009, 11, (15)3298-3301
  2. S.C.Mitchell; R.M.Nickson; E.R.Porter; W.F.Jackson; S.L.Preston; A.Q.Zhang. The fate of diphenyl sulphide, diphenyl sulphoxide and diphenyl sulphone in the rat. Drug Metabolism and Drug Interactions. 2000, 16, (3)191-206
  3. Sulfenylation agent for enolates of ketones, esters or carboxylic acid dianions: J. Am. Chem. Soc., 95, 6840 (1973); 98, 4887 (1976); Chem. Rev., 78, 363 (1978). Oxidation to the sulfoxide and thermal elimination can be used for the conversion of ketones to enones via thermal sulfoxide elimination, less frequently used than the selenoxide route (cf preceding entry), due to the higher temperatures required.
  4. Also sulfenylates organolithium reagents from, e.g. lithiated furans: J. Org. Chem., 46, 2473 (1981).
  5. Alkyl halides under phase-transfer conditions give alkyl phenyl sulfides in good yields,: Synth. Commun., 12, 595 (1982), and alkenyl halides in the presence of CuI give alkenyl sulfides: Chem. Lett., 769 (1989).
  6. For use as a catalyst in the photochemical cis-trans isomerization of olefins (used in the synthesis of the macrolide ricinelaidic acid lactone), see: Org. Synth. Coll., 7, 470 (1990).