3,4-Difluorophenol, 98%
3,4-Difluorophenol, 98%
3,4-Difluorophenol, 98%
Thermo Scientific Chemicals

3,4-Difluorophenol, 98%

CAS: 2713-33-9 | C6H4F2O | 130.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12596.22
also known as A12596-22
Price (USD)
520.65
Special offer
Online exclusive
Ends: 30-Jun-2026
612.00
Save 91.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
520.65
Special offer
Online exclusive
Ends: 30-Jun-2026
612.00
Save 91.35 (15%)
Each
Chemical Identifiers
CAS2713-33-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Cream to yellow to orange
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)32.0-39.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only