2-Fluoroaniline, 99%
2-Fluoroaniline, 99%
2-Fluoroaniline, 99%
Thermo Scientific Chemicals

2-Fluoroaniline, 99%

CAS: 348-54-9 | C6H6FN | 111.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12612.14
also known as A12612-14
Price (USD)
25.65
Online exclusive
28.50
Save 2.85
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
25.65
Online exclusive
28.50
Save 2.85
Each
Chemical Identifiers
CAS348-54-9
IUPAC Name2-fluoroaniline
Molecular FormulaC6H6FN
InChI KeyFTZQXOJYPFINKJ-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow to yellow to orange or pale pink
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5405-1.5435 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only