Polytetrafluoroethylene
Polytetrafluoroethylene
Polytetrafluoroethylene
Polytetrafluoroethylene
Thermo Scientific Chemicals

Polytetrafluoroethylene

CAS: 9002-84-0 | (C2F4)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12613.30
also known as A12613-30
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS9002-84-0
Molecular Formula(C2F4)n
SMILESFC(F)(-*)C(F)(F)-*
MDL NumberMFCD00084455
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Loss on Drying≤1% (UK-sourced material)
CommentMaterial sourced in UK and US
Melting Point327.5-337.5?C (US-sourced material)
FormPowder
PTFE may be used as a non stick coating. It can be used as engineering plastics. It is employed as lining reaction vessels, gaskets and pump packing, electrical insulator for high frequency applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only