Pyrrole, 98+%
Pyrrole, 98+%
Pyrrole, 98+%
Thermo Scientific Chemicals

Pyrrole, 98+%

CAS: 109-97-7 | C4H5N | 67.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12616.0B
also known as A12616-0B
Price (USD)
533.65
Online Exclusive
593.00
Save 59.35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
533.65
Online Exclusive
593.00
Save 59.35 (10%)
Each
Chemical Identifiers
CAS109-97-7
IUPAC Name1H-pyrrole
Molecular FormulaC4H5N
InChI KeyKAESVJOAVNADME-UHFFFAOYSA-N
SMILESN1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Assay (GC)≥98.0%
CommentMay darken on storage
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Pyrrole plays a major role in synthesis of drugs, spices, agrochemicals, dyes, photographic chemicals and perfumes. It plays an important role in the electropolymerisation of macroporous conducting polymer films. It acts as a catalyst for polymerization process; as a standard substance in chromatographic analysis; as corrosion inhibitors and preservatives and as solvents for resins and terpenes. It is utilized to study the hydrogen-bond mediated coupling of 1,2,3-triazole to pyrrole and in the preparation of 1-(4-Chloro-benzoyl)-pyrrole by reacting with 4-Chloro-benzoyl chloride. In Ciamician-Dennstedt rearrangement, It is used to prepare 3-chloropyridine by reacting with dichlorocarbene.

Solubility
Miscible with alcohol, ethyl ether, dilute acids and organic solvents. Immiscible with water.

Notes
Air, light and moisture sensitive. Store in cool place. Incompatible with strong acids, strong oxidizing agents, acid chlorides and acid anhydrides.
RUO – Research Use Only

General References:

  1. Williams, B. M.; Barone, V.; Pate, B. D.; Peralta, J. E. Gradient copolymers of thiophene and pyrrole for photovoltaics. Comput. Mater. Sci. 2015, 96, 69-71.
  2. Iyogun, A. A.; Kumar, M. R.; Freund, M. S. Chemically diverse sensor arrays based on electrochemically copolymerized pyrrole and styrene derivatives. Sens. Actuators, B 2015, 215, 510-517.