Thermo Scientific Chemicals

Methylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12619.06
5 g, Each
Thermo Scientific Chemicals

Methylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12619.06
5 g, Each
Quantity

Chemical Identifiers

CAS
993-13-5
IUPAC Name
methylphosphonic acid
Molecular Formula
CH5O3P
InChI Key
YACKEPLHDIMKIO-UHFFFAOYSA-N
SMILES
CP(O)(O)=O
Melting Point (clear melt)
103-109°C
Water Content (Karl Fischer Titration)
≤1.5%
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder.

Description

Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.

Solubility
Soluble in water and alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text