2,3-Dimethylpyrazine, 99%
2,3-Dimethylpyrazine, 99%
2,3-Dimethylpyrazine, 99%
Thermo Scientific Chemicals

2,3-Dimethylpyrazine, 99%

CAS: 5910-89-4 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A12628.09
also known as A12628-09
Price (USD)
38.65
Online Exclusive
42.90
Save 4.25 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
38.65
Online Exclusive
42.90
Save 4.25 (10%)
Each
Chemical Identifiers
CAS5910-89-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.5050-1.5080 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only