4-Bromobiphenyl, 98+%
4-Bromobiphenyl, 98+%
4-Bromobiphenyl, 98+%
Thermo Scientific Chemicals

4-Bromobiphenyl, 98+%

CAS: 92-66-0 | C12H9Br | 233.108 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12638.09
also known as A12638-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS92-66-0
IUPAC Name4-bromo-1,1'-biphenyl
Molecular FormulaC12H9Br
InChI KeyPKJBWOWQJHHAHG-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥98.0%
Melting Point (clear melt)87.0-93.0?C
Appearance (Color)White
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.

Solubility
Insoluble in water.

Notes
Sensitive to light. Store in cool dry place in tightly closed container. Avoid excessive heat and light. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. A Parkinson; and S Safe. Cytochrome P-450-mediated metabolism of biphenyl and the 4-halobiphenyls. Biochemical Pharmacology. 1982, 31(10), 1849-1856.
  2. Rusling JF, et al. Electrocatalytic reactions in organized assemblies: Part I. Reduction of 4-bromobiphenyl in cationic and non-ionic micelles. J. Electroanal. Chem. Interfac. Electrochem. 1988, 240(1), 201-216.