2-Ethylhexanoic acid, 99%
2-Ethylhexanoic acid, 99%
2-Ethylhexanoic acid, 99%
Thermo Scientific Chemicals

2-Ethylhexanoic acid, 99%

CAS: 149-57-5 | C8H16O2 | 144.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
100 mL
500 mL
2500 mL
Catalog number A12644.AE
also known as A12644-AE
Price (USD)
35.40
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
35.40
Each
Chemical Identifiers
CAS149-57-5
IUPAC Name2-ethylhexanoic acid
Molecular FormulaC8H16O2
InChI KeyOBETXYAYXDNJHR-UHFFFAOYNA-N
SMILESCCCCC(CC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4235-1.4265 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.

Solubility
Miscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Sun, T.; Qi, M.; Fu, R. Perhydroxylcucurbit[6]uril as a highly selective gas chromatographic stationary phase for analytes of wide-ranging polarity. J. Sep. Sci. 2015, 38 (5), 821-824.
  2. Mochalski, P.; Theurl, M.; Sponring, A.; Unterkofler, K.; Kirchmair, R.; Amann, A. Analysis of Volatile Organic Compounds Liberated and Metabolised by Human Umbilical Vein Endothelial Cells (HUVEC) In Vitro. Cell Biochem. Biophys. 2015, 71 (1), 323-329.