Thermo Scientific Chemicals

Sodium 1-butanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: A12654.22
100 g, Each
Thermo Scientific Chemicals

Sodium 1-butanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: A12654.22
100 g, Each
Quantity
Catalog number: A12654.22
also known as A12654-22
Price (USD)
Price: 486.00
Online price: 413.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2386-54-1
IUPAC Name
sodium butane-1-sulfonate
Molecular Formula
C4H9NaO3S
InChI Key
XQCHMGAOAWZUPI-UHFFFAOYSA-M
SMILES
[Na+].CCCCS([O-])(=O)=O
Appearance (Color)
White
Form
Powder
Assay (ex Sulphated Ash)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
<1.0%

Description

Sodium 1-butanesulfonate was used in the synthesis of high silicon content SAPO4-5. It was also used as a mobile phase to separate eight selenium compounds (selenite, selenate, selenocystine, selenourea, selenomethionine, selenoethionine, selenocystamine and trimethylselenonium ion). Ion-associating reagent for HPLC1 and reversed-phase chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-butanesulfonate was used in the synthesis of high silicon content SAPO4-5. It was also used as a mobile phase to separate eight selenium compounds (selenite, selenate, selenocystine, selenourea, selenomethionine, selenoethionine, selenocystamine and trimethylselenonium ion). Ion-associating reagent for HPLC1 and reversed-phase chromatography.

Solubility
Soluble in water (1-10%).

Notes
Sensitive to light. Store in cool dry place in tightly closed container. Avoid excessive heat and light. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text