(3-Phenylpropyl)triphenylphosphonium bromide, 97+%
(3-Phenylpropyl)triphenylphosphonium bromide, 97+%
(3-Phenylpropyl)triphenylphosphonium bromide, 97+%
Thermo Scientific Chemicals

(3-Phenylpropyl)triphenylphosphonium bromide, 97+%

CAS: 7484-37-9 | C27H26BrP | 461.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A12669.09
also known as A12669-09
Price (USD)
54.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
54.00
Each
Chemical Identifiers
CAS7484-37-9
IUPAC Nametriphenyl(3-phenylpropyl)phosphanium bromide
Molecular FormulaC27H26BrP
InChI KeyRPUZOJFXAPSSJD-UHFFFAOYSA-M
SMILES[Br-].C(CC1=CC=CC=C1)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Bromide)≥97.0 to ≤103.0%
Water Content (Karl Fischer Titration)≤2.5%
Appearance (Color)White to cream
FormPowder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only