4,4-Dimethyl-2-oxazoline, 98%
4,4-Dimethyl-2-oxazoline, 98%
4,4-Dimethyl-2-oxazoline, 98%
4,4-Dimethyl-2-oxazoline, 98%
Thermo Scientific Chemicals

4,4-Dimethyl-2-oxazoline, 98%

CAS: 30093-99-3 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12670.22
also known as A12670-22
Price (USD)
648.65
Online Exclusive
720.00
Save 71.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
648.65
Online Exclusive
720.00
Save 71.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS30093-99-3
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Refractive Index1.4170-1.4240 @ 20?C
Assay (GC)≥97.5%
4,4-Dimethyl-2-oxazoline is used in the fictionalization of a non-activated carbon-hydrogen bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Dimethyl-2-oxazoline is used in the fictionalization of a non-activated carbon-hydrogen bond.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Sean H. Wiedemann; Robert G. Bergman; and Jonathan A. Ellman. Rhodium-Catalyzed Direct C-H Addition of 4,4-Dimethyl-2-oxazoline to Alkenes. Org. Lett. 2004, 6(10), 1685-1687.
  2. G. Balavoine; J.C. Clinet. Cyclopalladated 2-t-butyl-4,4-dimethyl-2-oxazoline: its preparation, and use in the functionalisation of a non-activated carbon-hydrogen bond. Journal of Organometallic Chemistry. 1990, 390(3), c84-c88.
  3. For reviews of the chemistry of oxazolines, see: Chem. Rev., 71, 483 (1971); Angew. Chem. Int. Ed., 15, 270 (1976); Tetrahedron, 41, 837 (1985); 50, 2297 (1994).