4-Aminobenzoic acid, 99%
4-Aminobenzoic acid, 99%
4-Aminobenzoic acid, 99%
4-Aminobenzoic acid, 99%
Thermo Scientific Chemicals

4-Aminobenzoic acid, 99%

CAS: 150-13-0 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12673.30
also known as A12673-30
Price (USD)
56.00
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
56.00
Each
Add to cart
Chemical Identifiers
CAS150-13-0
IUPAC Name4-aminobenzoic acid
Molecular FormulaC7H7NO2
InChI KeyALYNCZNDIQEVRV-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
FormPowder
Assay (Silylated GC)≥98.5
Appearance (Color)White to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Aminobenzoic acid acts as a gastric acid secretion inhibitor. It is employed as an intermediate in the synthesis of folate (Vitamin BX), azo dyes and cross linking agents. It is also useful in biomedical sector for the preparation of local anesthetic and ointments. It is widely used in industries to make pharmaceutical products, flavors and preservatives. It is actively involved as a UV-blocking ingredient in sun tan cosmetics.

Solubility
Slightly soluble in water. Soluble in methanol and diethyl ether.

Notes
Air and light sensitive. Incompatible with ferric salts and oxidizing agents. May discolor on exposure to light and air. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Liu, B.-T.; Li, P.-S. Preparation and characterization of high-refractive-index polymer/inorganic hybrid films containing TiO2 nanoparticles prepared by 4-aminobenzoic acid. Surf. Coat. Technol. 2013, 231, 301-306.
  2. Campbell, J. L.; Blanc, J. C. Y. L.; Schneider, B. B. Probing Electrospray Ionization Dynamics Using Differential Mobility Spectrometry: The Curious Case of 4-Aminobenzoic Acid. Anal. Chem. 2012, 84 (18), 7857-7864.