Methyl thiophene-2-carboxylate, 97%
Methyl thiophene-2-carboxylate, 97%
Methyl thiophene-2-carboxylate, 97%
Thermo Scientific Chemicals

Methyl thiophene-2-carboxylate, 97%

CAS: 5380-42-7 | C6H6O2S | 142.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12677.22
also known as A12677-22
Price (USD)
538.65
Special offer
Online exclusive
Ends: 30-Jun-2026
633.00
Save 94.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
538.65
Special offer
Online exclusive
Ends: 30-Jun-2026
633.00
Save 94.35 (15%)
Each
Chemical Identifiers
CAS5380-42-7
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5400-1.5450 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only